Order Entry
Switzerland
Orders LinkContactUsLinkComponent
L(+)-Cysteine

L(+)-Cysteine

Bottle, 1 kg
Catalog # APOSOR0325-1KG
Irritant
Supplier:   Apollo Scientific
L(+)-Cysteine
Catalog # APOSOR0325-1KG
Supplier:  Apollo Scientific
CAS Number:  52-90-4
L(+)-Cysteine

Bottle, 1 kg

CAS Number:  52-90-4
Price
SFr. 113.00
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  H-Cys-OH, L-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid
  • CAS Number:52-90-4
  • Formula:HSCH₂CH(NH₂)CO₂H
  • MDL Number:MFCD00064306
  • Molecular Weight:121.16 g/mol
  • Melting Pt:240 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:200-158-2
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.