L(+)-Cysteine
Bottle, 100 g
Catalog # APOSOR0325-100G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: H-Cys-OH, L-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid
- CAS Number:52-90-4
- Formula:HSCH₂CH(NH₂)CO₂H
- MDL Number:MFCD00064306
- Molecular Weight:121.16 g/mol
- Melting Pt:240 °C (dec.)
- Storage Temperature:Ambient
- EINECS:200-158-2
Specifications
Specifications
- Pack type:Bottle
- Pk:100 g



