Order Entry
Switzerland
Orders LinkContactUsLinkComponent
4-Methylmorpholine

4-Methylmorpholine

Bottle, 100 mL
Catalog # APOSOR16938-100ML
Flammable
Corrosive
Irritant
Supplier:   Apollo Scientific
4-Methylmorpholine
Catalog # APOSOR16938-100ML
Supplier:  Apollo Scientific
CAS Number:  109-02-4
4-Methylmorpholine

Bottle, 100 mL

CAS Number:  109-02-4
Price
SFr. 36.40
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  NMM, N-Methylmorpholine
  • CAS Number:109-02-4
  • Formula:C₅H₁₁NO
  • MDL Number:MFCD00006175
  • Molecular Weight:101.15 g/mol
  • Boiling Pt:115…116 °C
  • Melting Pt:–66 °C
  • Density:0.92 g/cm³ (25 °C)
  • Flash Pt:14 °C
  • Storage Temperature:Ambient
  • EINECS:203-640-0
  • UN:2535
  • ADR:3+8,II
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mL

Specifications