Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Dog Recombinant Leptin

Dog Recombinant Leptin

1 mg
Catalog # APOSBITP1203-1MG
Supplier:   Apollo Scientific
Dog Recombinant Leptin
Catalog # APOSBITP1203-1MG
Supplier:  Apollo Scientific
CAS Number:  
Dog Recombinant Leptin

1 mg

CAS Number:  
Price
SFr. 3’030.00
Log in for availability
1 * 1 mg
Product Details & Documents
Specifications

Specifications

  • Pk:
    1 mg
  • Cat. No.:
    APOSBITP1203-1MG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    Leptin
  • Species:
    Dog

Specifications