Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Agar

Agar

Bottle, 5 kg
Catalog # APOSBIM1002-5KG
Irritant
Supplier:   Apollo Scientific
Agar
Catalog # APOSBIM1002-5KG
Supplier:  Apollo Scientific
CAS Number:  9002-18-0
Agar

Bottle, 5 kg

CAS Number:  9002-18-0
Price
650.00 SFr.
Log in for availability
1 * 5 kg
Product Details & Documents

A purified agar with a high gel strength and clarity for tissue culture and microbiological work. Minimal conc. of 5.0g/L to obtain solid gel.

Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Storage Temperature:Ambient
  • EINECS:232-658-1
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.