Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Methotrexate

Methotrexate

Bottle, 1 g
Catalog # APOSBIM0130-1G
Toxic
Health
Supplier:   Apollo Scientific
Methotrexate
Catalog # APOSBIM0130-1G
Supplier:  Apollo Scientific
CAS Number:  59-05-2
Methotrexate

Bottle, 1 g

CAS Number:  59-05-2
Price
SFr. 26.00
Log in for availability
1 * 1 g
Product Details & Documents

An antineoplastic agent which acts as an antimetaboliteof folic acid.

Synonyms:  (S)-2-(4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioic acid
  • CAS Number:59-05-2
  • Formula:C₂₀H₂₂N₈O₅
  • MDL Number:MFCD00150847
  • Molecular Weight:454.44 g/mol
  • Melting Pt:185…204 °C
  • Storage Temperature:Freezer
  • UN:1544
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.