Actinomycin D ≥99%
Bottle, 25 mg
Catalog # APOSBIA0175-25MG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Antineoplastic antibiotic
- Purity: ≥99%
- CAS Number:50-76-0
- Formula:C₆₂H₈₆N₁₂O₁₆
- MDL Number:MFCD00005033
- Storage Temperature:Freezer
- EINECS:200-063-6
- UN:3462
- ADR:6.1,II
Specifications
Specifications
- Pack type:Bottle
- Pk:25 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



