Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

α-Cyclodextrin

Bottle, 25 g
Catalog # APOSBIB6038-25G
Irritant
Supplier:   Apollo Scientific
α-Cyclodextrin
Catalog # APOSBIB6038-25G
Supplier:  Apollo Scientific
CAS Number:  10016-20-3
α-Cyclodextrin

Bottle, 25 g

CAS Number:  10016-20-3
Price
SFr. 34.70
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:10016-20-3
  • Formula:C₃₆H₆₀O₃₀
  • MDL Number:MFCD00078207
  • Molecular Weight:972.84 g/mol
  • Melting Pt:>278 °C
  • Melting Pt:>278 °C
  • Storage Temperature:Ambient
  • EINECS:233-007-4
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.