Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

α-Cyclodextrin

Bottle, 100 g
Catalog # APOSBIB6038-100G
Irritant
Supplier:   Apollo Scientific
α-Cyclodextrin
Catalog # APOSBIB6038-100G
Supplier:  Apollo Scientific
CAS Number:  10016-20-3
α-Cyclodextrin

Bottle, 100 g

CAS Number:  10016-20-3
Price
78.00 SFr.
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:10016-20-3
  • Formula:C₃₆H₆₀O₃₀
  • MDL Number:MFCD00078207
  • Molecular Weight:972.84 g/mol
  • Melting Pt:>278 °C
  • Melting Pt:>278 °C
  • Storage Temperature:Ambient
  • EINECS:233-007-4
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications