ContactUsLinkComponent
DT Water Vapour 3/a Measuring range: 3.0 - 60 lbs/mmcf (3 strokes, scale n=3) 1 * 10 items
Catalog # DRAE8103031
Supplier:  Dräger
DT Water Vapour 3/a Measuring range: 3.0 - 60 lbs/mmcf (3 strokes, scale n=3) 1 * 10 items
Catalog # DRAE8103031
Supplier:  Dräger