ContactUsLinkComponent
Inner chamber, reinforced - including 2 reinforced racks (max. total load 250 kg, max. load per rack 70 kg) 1 * 1 items
Catalog # BIND8012-1715
Supplier:  Binder
Inner chamber, reinforced - including 2 reinforced racks (max. total load 250 kg, max. load per rack 70 kg) 1 * 1 items
Catalog # BIND8012-1715
Supplier:  Binder
Frequently Bought Together