Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Dropping funnels, cylindrical, with double jacket

Dropping funnels, cylindrical, with double jacket

Dropping funnel with drilling 2,5 mm, 50 ml, 14/23 NS
Catalog # NEUB1596-20-0050
Dropping funnels, cylindrical, with double jacket
Catalog # NEUB1596-20-0050
Dropping funnels, cylindrical, with double jacket

Dropping funnel with drilling 2,5 mm, 50 ml, 14/23 NS

Price
SFr. 644.00
Log in for availability
1 * 1 ST
Product Details & Documents

Borosilicate glass 3.3, with PTFE tap.

  • Graduated
  • With drop counter
  • with GL 14 olives (PE)
  • With pressure compensation
Specifications
  • Socket size:
    14/23 NS
  • Capacity:
    50 ml
  • Description:
    Dropping funnel with drilling 2,5 mm
  • Seq:
    01