ContactUsLinkComponent
External dimensions WxDxH: ca. 985 x 751 x 1471 mm Internal dimensions WxDxH: ca. 745 x 650 x 1126 mm Slotted rail adjustment factor: 61,5 mm 2 steering castors with locks, 2 fixed castors, housing: galvanised, ø160 mm 2 x Norm5 solid shelves 1 * 1 items
Catalog # HUPF7504647
Supplier:  HUPFER METALLWERKE
External dimensions WxDxH: ca. 985 x 751 x 1471 mm Internal dimensions WxDxH: ca. 745 x 650 x 1126 mm Slotted rail adjustment factor: 61,5 mm 2 steering castors with locks, 2 fixed castors, housing: galvanised, ø160 mm 2 x Norm5 solid shelves 1 * 1 items
Catalog # HUPF7504647
Supplier:  HUPFER METALLWERKE
Frequently Bought Together