ContactUsLinkComponent
Rat Intrathecal Catheter (Alzet) Specifications: 10 cm of 28G PU (0.36 mm OD 0.18 mm ID) connected to 12.7 cm PU (0.84 mm OD 0.36 mm ID
Catalog # CRIV0007740
Supplier:  CHARLES RIVER
Rat Intrathecal Catheter (Alzet) Specifications: 10 cm of 28G PU (0.36 mm OD 0.18 mm ID) connected to 12.7 cm PU (0.84 mm OD 0.36 mm ID
Catalog # CRIV0007740
Supplier:  CHARLES RIVER
Frequently Bought Together