ContactUsLinkComponent
3 Adapter for storing sensors with 12 mm diameter and Pg13.5 thread in the HandyLab 7XX tubular Z541 1 * 1 items
Catalog # SCOI285205510
Supplier:  SI Analytics
3 Adapter for storing sensors with 12 mm diameter and Pg13.5 thread in the HandyLab 7XX tubular Z541 1 * 1 items
Catalog # SCOI285205510
Supplier:  SI Analytics