ContactUsLinkComponent
DAVIDS BIOTECHN. PEPTIDE PACKAGE (ONE PEPTIDE), SYNTHESIS OF 15MG PEPTIDE, PURITY: >70%, QUALITY CONTROL: HPLC & MS, CONJUGATION OF 5MG TO KLH FOR IMMUNIZATION, ALIQUOT OF THE FREE PEPTIDE (1-5MG
Catalog # XXXXP101
Supplier:  Chemicals
DAVIDS BIOTECHN. PEPTIDE PACKAGE (ONE PEPTIDE), SYNTHESIS OF 15MG PEPTIDE, PURITY: >70%, QUALITY CONTROL: HPLC & MS, CONJUGATION OF 5MG TO KLH FOR IMMUNIZATION, ALIQUOT OF THE FREE PEPTIDE (1-5MG
Catalog # XXXXP101
Supplier:  Chemicals
Frequently Bought Together