ContactUsLinkComponent
BioHazzardLogistic Box Dimensions aluminium box: outer: 455 x 340 x 375 mm dimension inner insulation: outer: 385 x 257 x 335 mm Inner : 290 x 168 x 240 mm weight: 3,68kg Sturdy Aluminium box with Insulation Outer Dimensions: 385 x 285 x 343 mm Inner 1 * 1 items
Catalog # DLTTB100018
Supplier:  DELTA T
BioHazzardLogistic Box Dimensions aluminium box: outer: 455 x 340 x 375 mm dimension inner insulation: outer: 385 x 257 x 335 mm Inner : 290 x 168 x 240 mm weight: 3,68kg Sturdy Aluminium box with Insulation Outer Dimensions: 385 x 285 x 343 mm Inner 1 * 1 items
Catalog # DLTTB100018
Supplier:  DELTA T
Frequently Bought Together