Orders LinkContactUsLinkComponent
Objectives, Panthera
CCIS® Plan achromatic objective UC PL 60×/0,8/S (WD=0,35 mm), 0,35 mm
Catalog # MOTI1101001705241
Supplier:   MOTIC
Objectives, Panthera
Catalog # MOTI1101001705241
Supplier:  MOTIC
Objectives, Panthera
Objectives, Panthera
CCIS® Plan achromatic objective UC PL 60×/0,8/S (WD=0,35 mm), 0,35 mm
Price
SFr. 223.00
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Magnification:
    60x/0,8
  • Description:
    CCIS® Plan achromatic objective UC PL 60×/0,8/S (WD=0,35 mm)
  • Seq:
    06
  • Working distance:
    0,35 mm