Orders LinkContactUsLinkComponent
Objectives, Panthera
Objectives, Panthera
CCIS® Plan achromatic objective UC PL 20×/0,45 (WD=0,8 mm), 0,8 mm
Catalog # MOTI1101001705221
Supplier:   MOTIC
Objectives, Panthera
Catalog # MOTI1101001705221
Supplier:  MOTIC
Objectives, Panthera
Objectives, Panthera
CCIS® Plan achromatic objective UC PL 20×/0,45 (WD=0,8 mm), 0,8 mm
Price
SFr. 148.00
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Magnification:
    20x/0,45
  • Description:
    CCIS® Plan achromatic objective UC PL 20×/0,45 (WD=0,8 mm)
  • Seq:
    04
  • Working distance:
    0,8 mm