ContactUsLinkComponent
Wavelength Calibration Standard for UV-Vis. 4% Holmium Oxide in 10% Perchloric Acid. 60ml Manufatured to NIST Std SRM-02034.Chem So lutions. 1 * 60 mL
Catalog # MICSPQA-HOX-A
Supplier:  MICROSOLV
undefined
Wavelength Calibration Standard for UV-Vis. 4% Holmium Oxide in 10% Perchloric Acid. 60ml Manufatured to NIST Std SRM-02034.Chem So lutions. 1 * 60 mL
Catalog # MICSPQA-HOX-A
Supplier:  MICROSOLV
Frequently Bought Together