ContactUsLinkComponent
Caps, screw top with Soft-Guard white silicone/ red PTFE septa, bonded in ribbed cap. Green, for use with 9-425mm, 12x32mm autosampl er vials. BASIK brand. 1,000/cs 1 * 1.000 items
Catalog # MICS9502S-E-10C-GM
Supplier:  MICROSOLV
undefined
Caps, screw top with Soft-Guard white silicone/ red PTFE septa, bonded in ribbed cap. Green, for use with 9-425mm, 12x32mm autosampl er vials. BASIK brand. 1,000/cs 1 * 1.000 items
Catalog # MICS9502S-E-10C-GM
Supplier:  MICROSOLV
Frequently Bought Together