ContactUsLinkComponent
Caps, screw top with Soft-Guard white silicone/ red PTFE septa, bonded in ribbed cap. Blue, for use with 9-425mm, 12x32mm autosample r vials. BASIK brand. 1,000/cs 1 * 1.000 items
Catalog # MICS9502S-E-10C-BM
Supplier:  MICROSOLV
undefined
Caps, screw top with Soft-Guard white silicone/ red PTFE septa, bonded in ribbed cap. Blue, for use with 9-425mm, 12x32mm autosample r vials. BASIK brand. 1,000/cs 1 * 1.000 items
Catalog # MICS9502S-E-10C-BM
Supplier:  MICROSOLV
Frequently Bought Together