Caps and septa, BASIK™
Snap cap and septa, pre-slit, Soft-Guard™, White silicone/red PTFE, Clear
Catalog # MICS9502C-E-30C
Supplier: MICROSOLV
Jump to:
Product Details & Documents
For use with 12×32 mm autosampler vials.
Specifications
- Septum:White silicone/red PTFE
- Description:Snap cap and septa, pre-slit, Soft-Guard™
- Seq:19
- Cap colour:Clear