Caps and septa, BASIK™
Snap cap and septa, non-slit, yellow, Soft-Guard™, White silicone/red PTFE, Yellow
Katalognr. MICS9502C-E-10Y-M
Anbieter: MICROSOLV
Direkt zu
Product Details & Documents
For use with 12×32 mm autosampler vials.
Technische Daten
- Septum:White silicone/red PTFE
- Description:Snap cap and septa, non-slit, yellow, Soft-Guard™
- Seq:14
- Cap colour:Yellow