ContactUsLinkComponent
The VWR Standard Lighted Tissue Flotation Bath helps to eliminate wrinkles and distortion during preparation of paraffin embedded tissue sections. The LED illuminated black background allows specimens to be easily seen and the digital microprocessor 1 * 1 items
Catalog # BOEI145702V-2
Supplier:  BOEKEL SCIENTIFIC
The VWR Standard Lighted Tissue Flotation Bath helps to eliminate wrinkles and distortion during preparation of paraffin embedded tissue sections. The LED illuminated black background allows specimens to be easily seen and the digital microprocessor 1 * 1 items
Catalog # BOEI145702V-2
Supplier:  BOEKEL SCIENTIFIC
Frequently Bought Together