Orders LinkContactUsLinkComponent
Drum faucets | Avantor
Drum faucets
Internal flame arrester, adjustable, 3/4"
Katalognr. 119-0240
Drum faucets
Katalognr. 119-0240
Drum faucets
Drum faucets
Internal flame arrester, adjustable, 3/4"
Preis
SFr. 93.10
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
These faucets can be simply placed on your drum for leakproof dispensing of flammable liquids. The brass construction with a polytetrafluoroethylene (PTFE) seal ensures excellent chemical resistance.
  • Safe and easy dispensing
  • FM approved
The faucets feature an internal flame arrester that prevents flashback of fire into the drum for added safety.
Technische Daten
  • Thread:
    3/4"
  • Description:
    Internal flame arrester, adjustable
  • Seq:
    02
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.