ContactUsLinkComponent
RF314DUAL WiFi Datalogger + 12-months FREE Cloud Application, Thermocouple with Sub-Min Connector (Dual External Temp) -270°C to +1300°C (-454°F to +2372°F) 1 * 1 items
Catalog # COMARF314DUAL
Supplier:  COMARK
RF314DUAL WiFi Datalogger + 12-months FREE Cloud Application, Thermocouple with Sub-Min Connector (Dual External Temp) -270°C to +1300°C (-454°F to +2372°F) 1 * 1 items
Catalog # COMARF314DUAL
Supplier:  COMARK
Frequently Bought Together