ContactUsLinkComponent
SpectraMax iD3 Multi-Mode Microplate Detection Platform. Including 12 months parts and labor warranty and 1 additional year of MRC Service Plan covering parts and labor. System Includes: Spectral Absorbance, Fluorescence, and Luminescence detection, 1 * 1 items
Catalog # 735-0371
Supplier:  Molecular Devices
SpectraMax iD3 Multi-Mode Microplate Detection Platform. Including 12 months parts and labor warranty and 1 additional year of MRC Service Plan covering parts and labor. System Includes: Spectral Absorbance, Fluorescence, and Luminescence detection, 1 * 1 items
Catalog # 735-0371
Supplier:  Molecular Devices
Frequently Bought Together