ContactUsLinkComponent
20L rectangular HDPE carboy only (100-415 mm PP cap removed and used to produce by LP/AC). Materials Supplied By: EP. Process: Particulate Cleaning & Gamma Irradiation (25-40 kGy). Packaging: Single cleanroom bag each bottle. Certification: C of A US 1 * 4 items
Catalog # NALGCENLV7L20PI-E
20L rectangular HDPE carboy only (100-415 mm PP cap removed and used to produce by LP/AC). Materials Supplied By: EP. Process: Particulate Cleaning & Gamma Irradiation (25-40 kGy). Packaging: Single cleanroom bag each bottle. Certification: C of A US 1 * 4 items
Catalog # NALGCENLV7L20PI-E