ContactUsLinkComponent
DESMOGLEIN 4 POLYCLONAL ANTIBODY, CY3 CONJUGATED, HOST: RABBIT, CROSS REACTIVE SPECIES: HUMAN, MOUSE, RAT, APPLICATION: IF(IHC-P), C ONCENTRATION: 1UG/UL, VOLUME: 100UL 1 * 100 µl
Catalog # BOSSBS-13744R-CY3
Supplier:  Bioss
DESMOGLEIN 4 POLYCLONAL ANTIBODY, CY3 CONJUGATED, HOST: RABBIT, CROSS REACTIVE SPECIES: HUMAN, MOUSE, RAT, APPLICATION: IF(IHC-P), C ONCENTRATION: 1UG/UL, VOLUME: 100UL 1 * 100 µl
Catalog # BOSSBS-13744R-CY3
Supplier:  Bioss