ContactUsLinkComponent
Vacuum model EXP1-15(CR) ClEANROOM w/ULPA 230V - INCLUDES ACCESSORIES Volt: 230V Herz: 50 Hz Watt: 1 Kw Ampere: 4.5 A Air flow: 224 m3/h Vacuum pressure: 1554 mmH2O Recovery capacity: Wet: 47L, Dry: 38L Dimensions: H97 x W 46 cm Weight: 37 kg N 1 * 1 items
Catalog # TIGV110254B
Supplier:  TIGER-VAC
Vacuum model EXP1-15(CR) ClEANROOM w/ULPA 230V - INCLUDES ACCESSORIES Volt: 230V Herz: 50 Hz Watt: 1 Kw Ampere: 4.5 A Air flow: 224 m3/h Vacuum pressure: 1554 mmH2O Recovery capacity: Wet: 47L, Dry: 38L Dimensions: H97 x W 46 cm Weight: 37 kg N 1 * 1 items
Catalog # TIGV110254B
Supplier:  TIGER-VAC
Frequently Bought Together