BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
Catalog # HACH25255061
Supplier: Hach
Frequently Bought Together
VWR® Facial Tissues
Supplier: VWR Collection
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
Measuring beakers with handle
Supplier: BURKLE
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Test strips, nitrite, Dosatest
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™ Xtra™
Supplier: KIMBERLY CLARK
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
pH/mV/°C meters, handheld, pH 3110 / 3310
Supplier: WTW
Formic acid 80% ≥80%, TECHNICAL
Supplier: VWR Chemicals
pH electrodes, SenTix® PLUS
Supplier: WTW
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
BIOTECTOR BASE REAGENT 1 * 20 L
Supplier: Hach