ContactUsLinkComponent
CUSTOM BOX OF 48 PHYTIP COLUMNS (500 µL VOLUME) CONTAINING 40 µL OF SEPHAROSE ANTI-EPO PER COLUMN FOR TECAN FREDOM EVO MCA 96- WELL PLATFORMS 1 * 1 KIT
Catalog # BITAPTM45-40-NB
Supplier:  BIOTAGE
CUSTOM BOX OF 48 PHYTIP COLUMNS (500 µL VOLUME) CONTAINING 40 µL OF SEPHAROSE ANTI-EPO PER COLUMN FOR TECAN FREDOM EVO MCA 96- WELL PLATFORMS 1 * 1 KIT
Catalog # BITAPTM45-40-NB
Supplier:  BIOTAGE
Frequently Bought Together