ContactUsLinkComponent
PROFOLDIN MicroMolar Histidine Assay Kit ( includes 750 µl 10 x C53 Dye and 50 µl of 10 mM Histidine. It is for measurement of 200 samples using 96-well plates. Cuvettes may also be used for measurements
Catalog # XXXXHIS200
Supplier:  Chemicals
PROFOLDIN MicroMolar Histidine Assay Kit ( includes 750 µl 10 x C53 Dye and 50 µl of 10 mM Histidine. It is for measurement of 200 samples using 96-well plates. Cuvettes may also be used for measurements
Catalog # XXXXHIS200
Supplier:  Chemicals
Frequently Bought Together