ContactUsLinkComponent
Burette 25:0.05ml CL.as with ns glass plug clear glass acc. to mohr,made of borosilicate glass 3.3 in compliance with din en iso 385 very good chemical resistance minimal thermal expansion high resistance to thermal shocks 1 * 2 items
Catalog # MARI2422011
Anbieter:  MARIENFELD
Burette 25:0.05ml CL.as with ns glass plug clear glass acc. to mohr,made of borosilicate glass 3.3 in compliance with din en iso 385 very good chemical resistance minimal thermal expansion high resistance to thermal shocks 1 * 2 items
Catalog # MARI2422011
Anbieter:  MARIENFELD
Frequently Bought Together