Calibration reference materials, alloys
Iron, three 50,8 mm pieces (780 °C), 1
Catalog # PERCN5320112
Supplier: PerkinElmer
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1
- Description:Iron, three 50,8 mm pieces (780 °C)
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



