Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Quick-RNA™ Plus Kits

Quick-RNA™ Plus Kits

Quick-RNA™ miniprep plus kit, 200 Preps, 1 KIT
Catalog # ZYMOR1058
Marketsource
Supplier:   Zymo Research
Quick-RNA™ Plus Kits
Catalog # ZYMOR1058
Supplier:  Zymo Research
Marketsource
CAS Number:  
Quick-RNA™ Plus Kits
Quick-RNA™ Plus Kits

Quick-RNA™ miniprep plus kit, 200 Preps, 1 KIT

CAS Number:  
Price
1 201.00 SFr.
Log in for availability
1 * 1 KIT
Product Details & Documents

RNA from any sample (cells, tissues, biological fluids and more).

  • DNA-free: Genomic DNA removal column and DNase I included
  • High quality: Ensure intact RNA with DNA/RNA Shield™ for sample storage at ambient temperatures
  • NGS-ready: RNA is ready for all downstream applications including next-gen sequencing, RT-qPCR, hybridization and more

The Quick-RNA™ miniprep plus kit is an innovative and versatile product designed for the easy, reliable, and rapid isolation of DNA-free RNA from cells, all tissue types, whole blood, and biological fluids. The provided DNA/RNA Shield™ stabilises samples, allowing them to be stored without the need for immediate freesing or processing for up to one month. Furthermore, DNA/RNA Shield™ inactivates RNases as well as microbial pathogens (viruses, bacteria and more). The procedure combines a unique buffer system with Zymo-Spin™ column technology to yield high quality total RNA (including small RNAs 17 to 200 nt).

Simply add DNA/RNA Shield™ and proteinase K to extract total RNA from any tissue, then purify the RNA using the Zymo-Spin™ column. The result is highly-concentrated, DNA-free RNA that is suitable for RT-PCR, hybridization, sequencing and more. In addition, the kit can be used for the enrichment of small and large RNAs in two separate fractions.

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    Quick-RNA™ miniprep plus kit, 200 Preps
  • Cat. No.:
    ZYMOR1058
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Target molecule:
    RNA
  • Method / Format:
    Column (silica-based)
  • Sample type:
    Cultured cells|Human/animal cells and tissues
  • Sample size:
    ≤1×10⁷ Cells or 50 mg Tissue
  • Sample capacity:
    100 µg
  • Yield:
    spin column, 100 µg RNA (binding capacity), ≥50 µl (elution volume)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.