Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Quick-RNA™ Plus Kits

Quick-RNA™ Plus Kits

Quick-RNA™ miniprep plus kit, 50 Preps, 1 KIT
Catalog # ZYMOR1057
Marketsource
Supplier:   Zymo Research
Quick-RNA™ Plus Kits
Catalog # ZYMOR1057
Supplier:  Zymo Research
Marketsource
CAS Number:  
Quick-RNA™ Plus Kits
Quick-RNA™ Plus Kits

Quick-RNA™ miniprep plus kit, 50 Preps, 1 KIT

CAS Number:  
Price
SFr. 377.00
Log in for availability
1 * 1 KIT
Product Details & Documents

RNA from any sample (cells, tissues, biological fluids and more).

  • DNA-free: Genomic DNA removal column and DNase I included
  • High quality: Ensure intact RNA with DNA/RNA Shield™ for sample storage at ambient temperatures
  • NGS-ready: RNA is ready for all downstream applications including next-gen sequencing, RT-qPCR, hybridization and more

The Quick-RNA™ miniprep plus kit is an innovative and versatile product designed for the easy, reliable, and rapid isolation of DNA-free RNA from cells, all tissue types, whole blood, and biological fluids. The provided DNA/RNA Shield™ stabilises samples, allowing them to be stored without the need for immediate freesing or processing for up to one month. Furthermore, DNA/RNA Shield™ inactivates RNases as well as microbial pathogens (viruses, bacteria and more). The procedure combines a unique buffer system with Zymo-Spin™ column technology to yield high quality total RNA (including small RNAs 17 to 200 nt).

Simply add DNA/RNA Shield™ and proteinase K to extract total RNA from any tissue, then purify the RNA using the Zymo-Spin™ column. The result is highly-concentrated, DNA-free RNA that is suitable for RT-PCR, hybridization, sequencing and more. In addition, the kit can be used for the enrichment of small and large RNAs in two separate fractions.

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    Quick-RNA™ miniprep plus kit, 50 Preps
  • Cat. No.:
    ZYMOR1057
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Target molecule:
    RNA
  • Method / Format:
    Column (silica-based)
  • Sample type:
    Cultured cells|Human/animal cells and tissues
  • Sample size:
    ≤1×10⁷ Cells or 50 mg Tissue
  • Sample capacity:
    100 µg
  • Yield:
    spin column, 100 µg RNA (binding capacity), ≥50 µl (elution volume)

Specifications