ContactUsLinkComponent
Features Modular Superflake ice machine, production up to 320 kg in 24 hours Hermetic compressor Air-cooled condenser Stainless steel auger and freezing cylinder (evaporator) Equipped with an electronic control board Ice breaker made in s 1 * 1 items
  SCOMMF47AS
 :  SCOTSMAN
Features Modular Superflake ice machine, production up to 320 kg in 24 hours Hermetic compressor Air-cooled condenser Stainless steel auger and freezing cylinder (evaporator) Equipped with an electronic control board Ice breaker made in s 1 * 1 items
  SCOMMF47AS
 :  SCOTSMAN
Frequently Bought Together