ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 10 MM IN DMSOGENZ-644282 IS A NON-CAMPTOTHECIN INHIBITOR OF TOPOISOMERASE I WIT H POTENTIAL ANTINEOPLASTIC ACTIVITY(IC50=1.2 NM).IC50 VALUE: 1.2 NM [1]TARGET: TOPOISOMERASE IIN VITRO: GENZ-644282 D 1 * 100 mg
Catalog # MDEXHY-16228-100MG
Supplier:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 10 MM IN DMSOGENZ-644282 IS A NON-CAMPTOTHECIN INHIBITOR OF TOPOISOMERASE I WIT H POTENTIAL ANTINEOPLASTIC ACTIVITY(IC50=1.2 NM).IC50 VALUE: 1.2 NM [1]TARGET: TOPOISOMERASE IIN VITRO: GENZ-644282 D 1 * 100 mg
Catalog # MDEXHY-16228-100MG
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together