LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO 15 MG/ML ETHANOL 5 MG/MLARN-509 IS A SELECTIVE AND COMPETITIVE ANDROGEN RE CEPTOR INHIBITOR WITH IC50 OF 16 NM, USEFUL FOR PROSTATE CANCER TREATMENT.IC50 VALUE: 16 NMTARGET: ANDROGEN RECEPTORA 1 * 50 mg
Catalog # MDEXHY-16060-50MG
Supplier: MEDCHEMEXPRESS
Frequently Bought Together