ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO 23 MG/MLSALINOMYCIN(PROCOXACIN) IS AN ANTIBACTERIAL AND COCCIDIOSTAT IONOP HORE THERAPEUTIC DRUG.IC50 VALUE:TARGET: ANTIBACTERIAL ANTICANCERSALINOMYCIN, A POLYETHER IONOPHORE ANTIBIOTIC ISOLA 1 * 5 mg
Catalog # MDEXHY-15597-5MG
Supplier:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO 23 MG/MLSALINOMYCIN(PROCOXACIN) IS AN ANTIBACTERIAL AND COCCIDIOSTAT IONOP HORE THERAPEUTIC DRUG.IC50 VALUE:TARGET: ANTIBACTERIAL ANTICANCERSALINOMYCIN, A POLYETHER IONOPHORE ANTIBIOTIC ISOLA 1 * 5 mg
Catalog # MDEXHY-15597-5MG
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together