ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 10 MM IN DMSO360A IS A STRONG AFFINITY AND SELECTIVITY INHIBITOR OF G-QUADRUPLE X STRUCTURES.IC50 VALUE:TARGET: G-QUADRUPLEXIN VITRO: WE FOUND A S-PHASE ACCUMULATION IN ATM-PROFICIENT, BUT NOT IN A 1 * 10 mg
Catalog # MDEXHY-15595A-10MG
Supplier:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 10 MM IN DMSO360A IS A STRONG AFFINITY AND SELECTIVITY INHIBITOR OF G-QUADRUPLE X STRUCTURES.IC50 VALUE:TARGET: G-QUADRUPLEXIN VITRO: WE FOUND A S-PHASE ACCUMULATION IN ATM-PROFICIENT, BUT NOT IN A 1 * 10 mg
Catalog # MDEXHY-15595A-10MG
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together