ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 10 MM IN DMSOTALTOBULIN (HTI-286 SPA-110) IS AN ANALOGUE OF HEMIASTERLIN POTE NT TUBULIN INHIBITOR ADCS CYTOTOXIN.IC50 VALUE:TARGET: TUBULININ VITRO: HTI-286 SIGNIFICANTLY INHIBITED PROLIFERATIO 1 * 50 mg
Catalog # MDEXHY-15584-50MG
Supplier:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 10 MM IN DMSOTALTOBULIN (HTI-286 SPA-110) IS AN ANALOGUE OF HEMIASTERLIN POTE NT TUBULIN INHIBITOR ADCS CYTOTOXIN.IC50 VALUE:TARGET: TUBULININ VITRO: HTI-286 SIGNIFICANTLY INHIBITED PROLIFERATIO 1 * 50 mg
Catalog # MDEXHY-15584-50MG
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together