ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 50 MM IN DMSOGSK126 IS A POTENT, HIGHLY SELECTIVE, S-ADENOSYL-METHIONINE-COMPET ITIVE, SMALL-MOLECULE INHIBITOR OF EZH2 METHYLTRANSFERASE WITH KI VALUE OF 0.5 NM.IC50 VALUE: 0.5 NM (KI)TARGET: EZH2 1 * 10 mg
Catalog # MDEXHY-13470-10MG
Supplier:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 50 MM IN DMSOGSK126 IS A POTENT, HIGHLY SELECTIVE, S-ADENOSYL-METHIONINE-COMPET ITIVE, SMALL-MOLECULE INHIBITOR OF EZH2 METHYLTRANSFERASE WITH KI VALUE OF 0.5 NM.IC50 VALUE: 0.5 NM (KI)TARGET: EZH2 1 * 10 mg
Catalog # MDEXHY-13470-10MG
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together