ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 10 MM IN DMSOKH-CB19 IS A POTENT AND HIGHLY SPECIFIC INHIBITOR OF THE CDC2-LIKE KINASE ISOFORMS 1 AND 4 (CLK1/CLK4).IC50 VALUE: 20 NM (CLK1) [1]TARGET: CLK1/4 INHIBITORIN VITRO: KH-CB19 SHOWED POT 1 * 5 mg
  MDEXHY-12828-5MG
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 10 MM IN DMSOKH-CB19 IS A POTENT AND HIGHLY SPECIFIC INHIBITOR OF THE CDC2-LIKE KINASE ISOFORMS 1 AND 4 (CLK1/CLK4).IC50 VALUE: 20 NM (CLK1) [1]TARGET: CLK1/4 INHIBITORIN VITRO: KH-CB19 SHOWED POT 1 * 5 mg
  MDEXHY-12828-5MG
Frequently Bought Together