ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 10 MM IN DMSOML216(CID-49852229) IS A POTENT INHIBITOR OF THE DNA UNWINDING ACT IVITY OF BLM HELICASE SHOWING SIMILAR IC50S OF 3.0 AND 0.97 ΜM FOR FULL LENGTH BLM AND BLM636–1298 RESPECTIVELY.IC50 1 * 50 mg
Catalog # MDEXHY-12342-50MG
Supplier:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 10 MM IN DMSOML216(CID-49852229) IS A POTENT INHIBITOR OF THE DNA UNWINDING ACT IVITY OF BLM HELICASE SHOWING SIMILAR IC50S OF 3.0 AND 0.97 ΜM FOR FULL LENGTH BLM AND BLM636–1298 RESPECTIVELY.IC50 1 * 50 mg
Catalog # MDEXHY-12342-50MG
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together