ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO ≥90MG/ML WATER MLN8054 IS A POTENT AND SELECTIV E INHIBITOR OF AURORA A WITH IC50 OF 4 NM EXHIBITS MORE THAN 40-FOLD SELECTIVE FOR AURORA A THAN AURORA B.IC50 VALUE 1 * 10 mg
Catalog # MDEXHY-10180-10MG
Supplier:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO ≥90MG/ML WATER MLN8054 IS A POTENT AND SELECTIV E INHIBITOR OF AURORA A WITH IC50 OF 4 NM EXHIBITS MORE THAN 40-FOLD SELECTIVE FOR AURORA A THAN AURORA B.IC50 VALUE 1 * 10 mg
Catalog # MDEXHY-10180-10MG
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together