ContactUsLinkComponent
Spectromelt® A50 50% lithium tetraborate / 50% lithium metaborate 1 * 1 kg
Catalog # 1.17057.1000
Supplier:  Merck
Spectromelt® A50 50% lithium tetraborate / 50% lithium metaborate 1 * 1 kg
Catalog # 1.17057.1000
Supplier:  Merck
Frequently Bought Together