ContactUsLinkComponent
11ml Aufschnappöffnung •Service temperature range of PFA is -328°F to 500°F (-200°C to 260°C) •Will fit most standard microcentrifuge models •RCF up to 10,000 g 1 * 1 ST
Catalog # SAVI200-915-50
Supplier:  SAVILLEX
11ml Aufschnappöffnung •Service temperature range of PFA is -328°F to 500°F (-200°C to 260°C) •Will fit most standard microcentrifuge models •RCF up to 10,000 g 1 * 1 ST
Catalog # SAVI200-915-50
Supplier:  SAVILLEX
Frequently Bought Together